Anti-ASGR1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200
Type:
Antigen: ASGR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77711-775
- 77711-776
- Antigen Symbol:
- ASGR1
- ASGR1
- Antigen Synonyms:
- ASGPR|Hepatic lectin H1|ASGPR1|ASGPR 1|ASGR1_HUMAN|HL-1|Asgr1|Asialoglycoprotein receptor 1|CLEC4H1|C type lectin domain family 4 member H1|C-type lectin domain family 4 member H1|ASGP-R 1
- ASGPR|Hepatic lectin H1|ASGPR1|ASGPR 1|ASGR1_HUMAN|HL-1|Asgr1|Asialoglycoprotein receptor 1|CLEC4H1|C type lectin domain family 4 member H1|C-type lectin domain family 4 member H1|ASGP-R 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P07306
- P07306
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Asialoglycoprotein receptor 1 protein (68-291AA)
- Recombinant Human Asialoglycoprotein receptor 1 protein (68-291AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together