Anti-AQP2 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: AQP2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77710-902
- 77710-903
- Antigen Symbol:
- AQP2
- AQP2
- Antigen Synonyms:
- WCH-CD|AQP CD|Collecting duct water channel protein|AQP2_HUMAN|Aquaporine 2|Aquaporin CD|AQP 2|Water channel protein for renal collecting duct|AQP-2|WCH CD|Water channel aquaporin 2|Aquaporin2|Aquaporin-CD|Aquaporin-2|AQP-CD|AQP2|Aquaporin 2 collecting duct|AQPCD|WCHCD|MGC34501|ADH water channel
- WCH-CD|AQP CD|Collecting duct water channel protein|AQP2_HUMAN|Aquaporine 2|Aquaporin CD|AQP 2|Water channel protein for renal collecting duct|AQP-2|WCH CD|Water channel aquaporin 2|Aquaporin2|Aquaporin-CD|Aquaporin-2|AQP-CD|AQP2|Aquaporin 2 collecting duct|AQPCD|WCHCD|MGC34501|ADH water channel
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P41181
- P41181
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Aquaporin-2 protein (225-271AA)
- Recombinant Human Aquaporin-2 protein (225-271AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together