Anti-ANXA3 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ANXA3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77711-536
- 77711-537
- Antigen Symbol:
- ANXA3
- ANXA3
- Antigen Synonyms:
- OTTHUMP00000219237|Placental anticoagulant protein III|ANXA 3|Annexin A3|ANX3|Lipocortin III|Annexin III|35-alpha calcimedin|OTTHUMP00000219238|PAP III|Calcimedin 35 alpha|PAP-III|2-cyclic phosphate 2-phosphohydrolase|Anxa3|Inositol 1 2 cyclic phosphate 2 phosphohydrolase|ANXA3_HUMAN|Annexin-3|OTTHUMP00000160701|Inositol 1
- OTTHUMP00000219237|Placental anticoagulant protein III|ANXA 3|Annexin A3|ANX3|Lipocortin III|Annexin III|35-alpha calcimedin|OTTHUMP00000219238|PAP III|Calcimedin 35 alpha|PAP-III|2-cyclic phosphate 2-phosphohydrolase|Anxa3|Inositol 1 2 cyclic phosphate 2 phosphohydrolase|ANXA3_HUMAN|Annexin-3|OTTHUMP00000160701|Inositol 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P12429
- P12429
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Annexin A3 protein (5-322AA)
- Recombinant Human Annexin A3 protein (5-322AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together