Anti-Alpl Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Alpl
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 77711-476
- 77711-475
- Antigen Symbol:
- Alpl
- Alpl
- Antigen Synonyms:
- Alpl|Akp2|EC 3.1.3.1|Alkaline phosphatase liver/bone/kidney isozyme|Alkaline phosphatase|Akp-2|TNSALP|AP-TNAP|tissue-nonspecific isozyme|Alkaline phosphatase 2
- Alpl|Akp2|EC 3.1.3.1|Alkaline phosphatase liver/bone/kidney isozyme|Alkaline phosphatase|Akp-2|TNSALP|AP-TNAP|tissue-nonspecific isozyme|Alkaline phosphatase 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P09242
- P09242
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Mouse Alkaline phosphatase, tissue-nonspecific isozyme protein (20-503AA)
- Recombinant Mouse Alkaline phosphatase, tissue-nonspecific isozyme protein (20-503AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together