Anti-AKT2 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: AKT2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77710-803
- 77710-804
- Antigen Symbol:
- AKT2
- AKT2
- Antigen Synonyms:
- v akt murine thymoma viral oncogene homolog 2|Protein kinase Akt 2|RAC-BETA|rac protein kinase beta|Protein kinase B beta|Akt2|PKBB|RAC-PK-beta|AKT2_HUMAN|murine thymoma viral (v-akt) homolog-2|V-AKT murine thymoma viral oncogene homolog 2|PKB|RACbeta|Murine thymoma viral (v-akt) homolog 2|PKB beta|RAC beta|RAC-beta serine/threonine-protein kinase|Oncogene AKT2 protein kinase B beta|PRKBB|PKBBETA|HIHGHH|Protein kinase Akt-2
- v akt murine thymoma viral oncogene homolog 2|Protein kinase Akt 2|RAC-BETA|rac protein kinase beta|Protein kinase B beta|Akt2|PKBB|RAC-PK-beta|AKT2_HUMAN|murine thymoma viral (v-akt) homolog-2|V-AKT murine thymoma viral oncogene homolog 2|PKB|RACbeta|Murine thymoma viral (v-akt) homolog 2|PKB beta|RAC beta|RAC-beta serine/threonine-protein kinase|Oncogene AKT2 protein kinase B beta|PRKBB|PKBBETA|HIHGHH|Protein kinase Akt-2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P31751
- P31751
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human RAC-beta serine/threonine-protein kinase protein (2-481AA)
- Recombinant Human RAC-beta serine/threonine-protein kinase protein (2-481AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together