Anti-ADK Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, WB:1:200-1:1000
Type:
Antigen: ADK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77711-554
- 77711-553
- Antigen Symbol:
- ADK
- ADK
- Antigen Synonyms:
- AI255373|OTTHUMP00000019865|MGC6593|Adenosine 5''-phosphotransferase|ADK_HUMAN|AK|adk|AI987814|Adenosine kinase|2310026J05Rik|OTTHUMP00000019864|5033405D03Rik
- AI255373|OTTHUMP00000019865|MGC6593|Adenosine 5''-phosphotransferase|ADK_HUMAN|AK|adk|AI987814|Adenosine kinase|2310026J05Rik|OTTHUMP00000019864|5033405D03Rik
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P55263
- P55263
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human ADK
- Fusion protein of Human ADK
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together