Anti-TLK2 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TLK2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77716-609
- 77716-610
- Antigen Symbol:
- TLK2
- TLK2
- Antigen Synonyms:
- Serine/threonine protein kinase tousled like 2|PKU-alpha|Serine/threonine-protein kinase tousled-like 2|Serine/threonine kinase|MGC44450|PKU alpha|Tousled like kinase 2|TLK2_HUMAN|TLK 2|tlk2|Tousled-like kinase 2
- Serine/threonine protein kinase tousled like 2|PKU-alpha|Serine/threonine-protein kinase tousled-like 2|Serine/threonine kinase|MGC44450|PKU alpha|Tousled like kinase 2|TLK2_HUMAN|TLK 2|tlk2|Tousled-like kinase 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86UE8
- Q86UE8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Serine/threonine-protein kinase tousled-like 2 protein (23-234AA)
- Recombinant Human Serine/threonine-protein kinase tousled-like 2 protein (23-234AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together