Anti-CAB39L Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:50-1:200
Type:
Antigen: CAB39L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77716-378
- 77716-379
- Antigen Symbol:
- CAB39L
- CAB39L
- Antigen Synonyms:
- FLJ12577|U937-associated antigen|CAB39-LIKE PROTEIN|MO2L|sarcoma antigen NY-SAR-79|MO25-BETA|RP11-103J18.3|Cab39l|Calcium-binding protein 39-like|CB39L_HUMAN|Mo25-like protein|bA103J18.3|Antigen MLAA-34
- FLJ12577|U937-associated antigen|CAB39-LIKE PROTEIN|MO2L|sarcoma antigen NY-SAR-79|MO25-BETA|RP11-103J18.3|Cab39l|Calcium-binding protein 39-like|CB39L_HUMAN|Mo25-like protein|bA103J18.3|Antigen MLAA-34
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H9S4
- Q9H9S4
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human CAB39L
- Fusion protein of Human CAB39L
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together