Anti-IL31RA Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, IHC:1:25-1:100
Type:
Antigen: IL31RA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77716-357
- 77716-356
- Antigen Symbol:
- IL31RA
- IL31RA
- Antigen Synonyms:
- CRL 3|MGC125346|IL 31 receptor subunit alpha|HGLM-R|HGLM R|GLM-R|CRL|IL-31R subunit alpha|Gp130-like receptor|IL31 receptor subunit alpha|GP130 like receptor|Class I cytokine receptor|IL 31R subunit alpha|Interleukin-31 receptor subunit alpha|Gp130-like monocyte receptor|IL31R_HUMAN|IL-31RA|Soluble type I cytokine receptor CRL3|IL31RA|IL31R subunit alpha|IL-31R-alpha|PRO21384|PLCA2|Cytokine receptor-like 3|CRL3|GLMR|GLM R|HGLMR|IL 31RA|GPL|Cytokine receptor like 3|PLCA 2|Interleukin 31 receptor A|Gp130 like monocyte receptor|interleukin 31 receptor subunit alpha|IL-31 receptor subunit alpha|ZcytoR17
- CRL 3|MGC125346|IL 31 receptor subunit alpha|HGLM-R|HGLM R|GLM-R|CRL|IL-31R subunit alpha|Gp130-like receptor|IL31 receptor subunit alpha|GP130 like receptor|Class I cytokine receptor|IL 31R subunit alpha|Interleukin-31 receptor subunit alpha|Gp130-like monocyte receptor|IL31R_HUMAN|IL-31RA|Soluble type I cytokine receptor CRL3|IL31RA|IL31R subunit alpha|IL-31R-alpha|PRO21384|PLCA2|Cytokine receptor-like 3|CRL3|GLMR|GLM R|HGLMR|IL 31RA|GPL|Cytokine receptor like 3|PLCA 2|Interleukin 31 receptor A|Gp130 like monocyte receptor|interleukin 31 receptor subunit alpha|IL-31 receptor subunit alpha|ZcytoR17
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NI17
- Q8NI17
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human IL31RA
- Fusion protein of Human IL31RA
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together