Anti-DNAJB1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, IHC:1:25-1:100
Type:
Antigen: DNAJB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77716-323
- 77716-322
- Antigen Symbol:
- DNAJB1
- DNAJB1
- Antigen Synonyms:
- HDJ 1|Human DnaJ protein 1|DnaJ protein homolog 1|DnaJ homolog subfamily B member 1|Radial spoke 16 homolog B|Heat shock protein 40|HDJ-1|DNAJ 1|RSPH16B|HDJ1|Dnajb1|DNAJB 1|DnaJ heat shock protein family (Hsp40) member B1|HSPF1|DNAJB1 protein|DnaJ (Hsp40) homolog subfmaily B member 1|DNAJ B1|DNJB1_HUMAN|Heat shock 40 kDa protein 1|HSPF 1|Heat shock 40kD protein 1|HSP40|DNAJ1|Sis1|Hsp 40
- HDJ 1|Human DnaJ protein 1|DnaJ protein homolog 1|DnaJ homolog subfamily B member 1|Radial spoke 16 homolog B|Heat shock protein 40|HDJ-1|DNAJ 1|RSPH16B|HDJ1|Dnajb1|DNAJB 1|DnaJ heat shock protein family (Hsp40) member B1|HSPF1|DNAJB1 protein|DnaJ (Hsp40) homolog subfmaily B member 1|DNAJ B1|DNJB1_HUMAN|Heat shock 40 kDa protein 1|HSPF 1|Heat shock 40kD protein 1|HSP40|DNAJ1|Sis1|Hsp 40
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P25685
- P25685
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human DNAJB1
- Fusion protein of Human DNAJB1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together