Anti-ASPA Rabbit Polyclonal Antibody
New Product
ELISA:1:2000-1:10000, WB:1:1000-1:5000, IHC:1:100-1:300
Type:
Antigen: ASPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77716-189
- 77716-190
- Antigen Symbol:
- ASPA
- ASPA
- Antigen Synonyms:
- RP23-213I10.1|NUR 7|NUR7|ACY2_HUMAN|Aminoacylase 2|ACY-2|ACY 2|Aspartoacylase (aminoacylase 2|ASPA|OTTMUSP00000006437|ACY2|ASP|Aminoacylase2|Aspartoacylase|Small lethargic|Aspartoacylase (Canavan disease)|Aminoacylase-2|Canavan disease)
- RP23-213I10.1|NUR 7|NUR7|ACY2_HUMAN|Aminoacylase 2|ACY-2|ACY 2|Aspartoacylase (aminoacylase 2|ASPA|OTTMUSP00000006437|ACY2|ASP|Aminoacylase2|Aspartoacylase|Small lethargic|Aspartoacylase (Canavan disease)|Aminoacylase-2|Canavan disease)
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P45381
- P45381
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human ASPA
- Fusion protein of Human ASPA
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together