Anti-CASP3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:10000, WB:1:200-1:1000, IHC:1:25-1:100
Type:
Antigen: CASP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77716-029
- 77716-028
- Antigen Symbol:
- CASP3
- CASP3
- Antigen Synonyms:
- OTTHUMP00000165052|Caspase-3 subunit p12|CASP3_HUMAN|apoptosis-related cysteine peptidase|Procaspase3|A830040C14Rik|OTTHUMP00000165054|Yama|Protein Yama|OTTHUMP00000165053|Cysteine protease CPP32|Casp3a|LICE|CPP 32|Caspase3|CASP-3|PARP cleavage protease|CASP3|CPP32|Apopain|apoptosis-related cysteine protease|CASP 3|EC 3.4.22.56|apoptosis-related cysteine protease a|CC3|Yama protein|SCA1|CPP32B|mldy|SCA-1|SCA 1|CPP-32|SREBP cleavage activity 1|ICE3|Caspase 3
- OTTHUMP00000165052|Caspase-3 subunit p12|CASP3_HUMAN|apoptosis-related cysteine peptidase|Procaspase3|A830040C14Rik|OTTHUMP00000165054|Yama|Protein Yama|OTTHUMP00000165053|Cysteine protease CPP32|Casp3a|LICE|CPP 32|Caspase3|CASP-3|PARP cleavage protease|CASP3|CPP32|Apopain|apoptosis-related cysteine protease|CASP 3|EC 3.4.22.56|apoptosis-related cysteine protease a|CC3|Yama protein|SCA1|CPP32B|mldy|SCA-1|SCA 1|CPP-32|SREBP cleavage activity 1|ICE3|Caspase 3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P42574
- P42574
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human CASP3 (active)
- Synthetic peptide of Human CASP3 (active)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together