Anti-SPARC Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:25-1:100
Type:
Antigen: SPARC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77716-006
- 77716-005
- Antigen Symbol:
- SPARC
- SPARC
- Antigen Synonyms:
- Cysteine rich protein|Basement membrane protein 40|AA517111|Secreted protein acidic cysteine rich (osteonectin)|Secreted protein acidic cysteine rich|Secreted protein acidic and cysteine rich|Basement-membrane protein 40|BM 40|OI17|ON|Secreted acidic cystein rich glycoprotein|SPARC|SPRC|BM40|Osteonectin|SPRC_HUMAN|BM-40|MGC128090|hm:zeh0062|Secreted protein acidic and rich in cysteine
- Cysteine rich protein|Basement membrane protein 40|AA517111|Secreted protein acidic cysteine rich (osteonectin)|Secreted protein acidic cysteine rich|Secreted protein acidic and cysteine rich|Basement-membrane protein 40|BM 40|OI17|ON|Secreted acidic cystein rich glycoprotein|SPARC|SPRC|BM40|Osteonectin|SPRC_HUMAN|BM-40|MGC128090|hm:zeh0062|Secreted protein acidic and rich in cysteine
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P09486
- P09486
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human SPARC
- Fusion protein of Human SPARC
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together