Anti-ATP1B2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, IHC:1:25-1:100
Type:
Antigen: ATP1B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77715-955
- 77715-954
- Antigen Symbol:
- ATP1B2
- ATP1B2
- Antigen Synonyms:
- AMOG|AT1B2_HUMAN|Sodium/potassium dependent ATPase subunit beta 2|Sodium/potassium-transporting ATPase subunit beta-2|sodium/potassium dependent ATPase beta 2 subunit|sodium pump subunit beta 2|sodium/potassium transporting ATPase subunit beta 2|Sodium/potassium-dependent ATPase subunit beta-2|Na+/K+ -ATPase beta 2 subunit|MGC93648|RATATPB2S|Na|adhesion molecule on glia|ATPB2|sodium/potassium transporting ATPase beta 2 chain|AT1B2|ATPB2S|sodium potassium ATPase subunit beta 2 (non-catalytic)|ATP1B2|ATPase Na+/K+ transporting beta 2 polypeptide|K ATPase beta 2 polypeptide
- AMOG|AT1B2_HUMAN|Sodium/potassium dependent ATPase subunit beta 2|Sodium/potassium-transporting ATPase subunit beta-2|sodium/potassium dependent ATPase beta 2 subunit|sodium pump subunit beta 2|sodium/potassium transporting ATPase subunit beta 2|Sodium/potassium-dependent ATPase subunit beta-2|Na+/K+ -ATPase beta 2 subunit|MGC93648|RATATPB2S|Na|adhesion molecule on glia|ATPB2|sodium/potassium transporting ATPase beta 2 chain|AT1B2|ATPB2S|sodium potassium ATPase subunit beta 2 (non-catalytic)|ATP1B2|ATPase Na+/K+ transporting beta 2 polypeptide|K ATPase beta 2 polypeptide
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P14415
- P14415
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human ATP1B2
- Fusion protein of Human ATP1B2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together