Anti-HDAC2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:5000, IHC:1:25-1:100
Type:
Antigen: HDAC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77715-820
- 77715-821
- Antigen Symbol:
- HDAC2
- HDAC2
- Antigen Synonyms:
- HD 2|Histone deacetylase 2|Histone deacetylase 2 (HD2)|HD2|D10Wsu179e|Yy1bp|HDAC2_HUMAN|OTTHUMP00000227078|transcriptional regulator homolog RPD3|YY1 associated factor 1|YAF1|YY1 transcription factor binding protein|Hdac2|RPD3|OTTHUMP00000227077|OTTHUMP00000017046|HDAC 2
- HD 2|Histone deacetylase 2|Histone deacetylase 2 (HD2)|HD2|D10Wsu179e|Yy1bp|HDAC2_HUMAN|OTTHUMP00000227078|transcriptional regulator homolog RPD3|YY1 associated factor 1|YAF1|YY1 transcription factor binding protein|Hdac2|RPD3|OTTHUMP00000227077|OTTHUMP00000017046|HDAC 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92769
- Q92769
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human HDAC2
- Synthetic peptide of Human HDAC2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together