Anti-MAP4 Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:2000, IHC:1:15-1:50
Type:
Antigen: MAP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77715-688
- 77715-689
- Antigen Symbol:
- MAP4
- MAP4
- Antigen Synonyms:
- OTTHUMP00000210730|OTTHUMP00000210727|MGC8617|Microtubule-associated protein 4|OTTHUMP00000210725|MAP-4|Microtubule associated protein 4|OTTHUMP00000210723|MAP4_HUMAN|DKFZp779A1753|MAP4
- OTTHUMP00000210730|OTTHUMP00000210727|MGC8617|Microtubule-associated protein 4|OTTHUMP00000210725|MAP-4|Microtubule associated protein 4|OTTHUMP00000210723|MAP4_HUMAN|DKFZp779A1753|MAP4
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P27816
- P27816
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human MAP4
- Fusion protein of Human MAP4
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together