Anti-SSTR5 Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:5000, WB:1:200-1:1000, IHC:1:25-1:100
Type:
Antigen: SSTR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77715-623
- 77715-622
- Antigen Symbol:
- SSTR5
- SSTR5
- Antigen Synonyms:
- SS-5-R|Somatostatin receptor type 5|SS5-R|Somatostatin receptor subtype 5|body|SS 5R|SSR5_HUMAN|SSTR 5|SST R5|SS5R|SS5 R|Sstr5
- SS-5-R|Somatostatin receptor type 5|SS5-R|Somatostatin receptor subtype 5|body|SS 5R|SSR5_HUMAN|SSTR 5|SST R5|SS5R|SS5 R|Sstr5
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P35346
- P35346
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human SSTR5
- Synthetic peptide of Human SSTR5
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together