Anti-BRD4 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:500-1:2000
Type:
Antigen: BRD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77715-485
- 77715-484
- Antigen Symbol:
- BRD4
- BRD4
- Antigen Synonyms:
- bromodomain containing protein 4|Bromodomain-containing protein 4|Brd4|Mitotic chromosome-associated protein|MCAP|HUNKI|Protein HUNK1|BRD4-NUT fusion oncoprotein|HUNK1|BRD4_HUMAN|CAP|chromosome associated protein|BRD4-NUT FUSION|Bromodomain containing 4
- bromodomain containing protein 4|Bromodomain-containing protein 4|Brd4|Mitotic chromosome-associated protein|MCAP|HUNKI|Protein HUNK1|BRD4-NUT fusion oncoprotein|HUNK1|BRD4_HUMAN|CAP|chromosome associated protein|BRD4-NUT FUSION|Bromodomain containing 4
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60885
- O60885
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human BRD4
- Synthetic peptide of Human BRD4
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together