Anti-ARTN Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
IHC:1:50-300, ELISA:1:10000-20000
Type:
Antigen: ARTN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77705-375
- 77715-384
- Antigen Symbol:
- ARTN
- ARTN
- Antigen Synonyms:
- Artemin|Enovin|NBN|Artn|EVN|ARTN_HUMAN|Neublastin|Neurotrophic factor
- Artemin|Enovin|NBN|Artn|EVN|ARTN_HUMAN|Neublastin|Neurotrophic factor
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5T4W7
- Q5T4W7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide from Human protein at AA range: 101-150
- Synthetic peptide from Human protein at AA range: 101-150
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.4.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together