Order Entry
United States
ContactUsLinkComponent
Anti-CKMT1A Rabbit Polyclonal Antibody
Anti-CKMT1A Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-CKMT1A Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77715-383
  • 77715-382
  • Antigen Symbol:
  • CKMT1A
  • CKMT1A
  • Antigen Synonyms:
  • Acidic-type mitochondrial creatine kinase|Creatine kinase mitochondrial 1 (ubiquitous)|Mia-CK|KCRU_HUMAN|U-MtCK|Creatine kinase U-type|CKMT|mitochondrial|Ubiquitous mitochondrial creatine kinase|Creatine kinase mitochondrial 1B|UMTCK|CKMT1B|Creatine kinase U type mitochondrial|CKMT1
  • Acidic-type mitochondrial creatine kinase|Creatine kinase mitochondrial 1 (ubiquitous)|Mia-CK|KCRU_HUMAN|U-MtCK|Creatine kinase U-type|CKMT|mitochondrial|Ubiquitous mitochondrial creatine kinase|Creatine kinase mitochondrial 1B|UMTCK|CKMT1B|Creatine kinase U type mitochondrial|CKMT1
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P12532
  • P12532
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Synthetic peptide of Human CKMT1A/CKMT1B
  • Synthetic peptide of Human CKMT1A/CKMT1B
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together