Anti-ITPR2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:10000, IHC:1:50-1:200
Type:
Antigen: ITPR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77715-375
- 77715-376
- Antigen Symbol:
- ITPR2
- ITPR2
- Antigen Synonyms:
- Type 2 inositol 145 trisphosphate receptor|Inositol 145 trisphosphate receptor type 2|5-trisphosphate receptor|IP3 R2|ITPR 2|IP3 receptor|IP3 receptor isoform 2|IP3R 2|InsP3R2|ITPR2_HUMAN|4|Itpr2|InsP3 R2|Inositol 1,4,5-trisphosphate receptor type 2|IP3R2|Type 2 InsP3 receptor|Inositol 1 4 5 trisphosphate receptor type 2|Type 2 inositol 1 4 5 trisphosphate receptor|Type 2 inositol 1
- Type 2 inositol 145 trisphosphate receptor|Inositol 145 trisphosphate receptor type 2|5-trisphosphate receptor|IP3 R2|ITPR 2|IP3 receptor|IP3 receptor isoform 2|IP3R 2|InsP3R2|ITPR2_HUMAN|4|Itpr2|InsP3 R2|Inositol 1,4,5-trisphosphate receptor type 2|IP3R2|Type 2 InsP3 receptor|Inositol 1 4 5 trisphosphate receptor type 2|Type 2 inositol 1 4 5 trisphosphate receptor|Type 2 inositol 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14571
- Q14571
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human ITPR2
- Synthetic peptide of Human ITPR2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together