Anti-SIGLEC6 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:5000, IHC:1:50-1:200
Type:
Antigen: SIGLEC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77715-212
- 77715-211
- Antigen Symbol:
- SIGLEC6
- SIGLEC6
- Antigen Synonyms:
- CDw327|CD33L|SIGLEC6|CD327|Sialic acid-binding Ig-like lectin 6|SIGL6_HUMAN|Obesity-binding protein 1|OBBP1|CD33L2|CD33 antigen-like 1|CD33L1|OB-BP1|Siglec-6
- CDw327|CD33L|SIGLEC6|CD327|Sialic acid-binding Ig-like lectin 6|SIGL6_HUMAN|Obesity-binding protein 1|OBBP1|CD33L2|CD33 antigen-like 1|CD33L1|OB-BP1|Siglec-6
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43699
- O43699
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human SIGLEC6
- Synthetic peptide of Human SIGLEC6
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together