Anti-RYR1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, WB:1:200-1:1000
Type:
Antigen: RYR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77715-104
- 77715-103
- Antigen Symbol:
- RYR1
- RYR1
- Antigen Synonyms:
- Skeletal muscle-type ryanodine receptor|RYR|RYR1_HUMAN|Type 1 ryanodine receptor|CCO|RYR-1|Protein phosphatase 1 regulatory subunit 137|Skeletal muscle ryanodine receptor|SKRR|RYDR|sarcoplasmic reticulum calcium release channel|Skeletal muscle calcium release channel|MHS1|Central core disease of muscle|Ryanodine receptor 1|RyR1|MHS|PPP1R137
- Skeletal muscle-type ryanodine receptor|RYR|RYR1_HUMAN|Type 1 ryanodine receptor|CCO|RYR-1|Protein phosphatase 1 regulatory subunit 137|Skeletal muscle ryanodine receptor|SKRR|RYDR|sarcoplasmic reticulum calcium release channel|Skeletal muscle calcium release channel|MHS1|Central core disease of muscle|Ryanodine receptor 1|RyR1|MHS|PPP1R137
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P21817
- P21817
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human RYR1
- Synthetic peptide of Human RYR1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together