Anti-PRDX2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:100-1:300
Type:
Antigen: PRDX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77715-060
- 77715-059
- Antigen Symbol:
- PRDX2
- PRDX2
- Antigen Synonyms:
- Thiol Specific Antioxidant 1|Thioredoxin-dependent peroxide reductase 1|TPX1|PRDX2|TSA|PRX2|Torin|Thiol-specific antioxidant protein|Natural killer cell enhancing factor B|TDPX1|Peroxiredoxin-2|MGC4104|Natural Killer Enhancing Factor B|PRXII|PRDX 2|Epididymis secretory sperm binding protein Li 2a|NKEF-B|Natural killer cell-enhancing factor B|PRDX2_HUMAN|Thiol specific antioxidant protein|Thioredoxin peroxidase 1|HEL S 2a|Thioredoxin Dependent Peroxide Reductase 1|PrP|NKEF B|Peroxiredoxin 2|NKEFB|PTX1
- Thiol Specific Antioxidant 1|Thioredoxin-dependent peroxide reductase 1|TPX1|PRDX2|TSA|PRX2|Torin|Thiol-specific antioxidant protein|Natural killer cell enhancing factor B|TDPX1|Peroxiredoxin-2|MGC4104|Natural Killer Enhancing Factor B|PRXII|PRDX 2|Epididymis secretory sperm binding protein Li 2a|NKEF-B|Natural killer cell-enhancing factor B|PRDX2_HUMAN|Thiol specific antioxidant protein|Thioredoxin peroxidase 1|HEL S 2a|Thioredoxin Dependent Peroxide Reductase 1|PrP|NKEF B|Peroxiredoxin 2|NKEFB|PTX1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P32119
- P32119
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human PRDX2
- Synthetic peptide of Human PRDX2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together