Anti-XRCC6 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: XRCC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77705-368
- 77714-984
- Antigen Symbol:
- XRCC6
- XRCC6
- Antigen Synonyms:
- CTC box binding factor 75 kDa subunit|Kup70|ATP dependent DNA helicase II 70 kDa subunit|DNA repair protein XRCC6|XRCC 6|Ku 70|Ku autoantigen, 70kDa|Thyroid-lupus autoantigen|70 kDa subunit of Ku antigen|Ku autoantigen p70 subunit|XRCC6_HUMAN|TLAA|X-ray repair complementing defective repair in Chinese hamster cells 6|X ray repair complementing defective repair in Chinese hamster cells 6|Thyroid autoantigen 70kD (Ku antigen)|Ku70 DNA binding component of DNA-dependent proteinkinase complex (thyroid autoantigen 70 kDa|Thyroid lupus autoantigen|Xrcc6|CTC75|CTCBF|X-ray repair cross-complementing protein 6|ATP dependent DNA helicase 2 subunit 1|Ku70|CTC box-binding factor 75 kDa subunit|Thyroid lupus autoantigen p70|ATP-dependent DNA helicase 2 subunit 1|ML8|5''-dRP lyase Ku70|Lupus Ku autoantigen protein p70|Thyroid autoantigen|G22P1|5''-deoxyribose-5-phosphate lyase Ku70|ATP-dependent DNA helicase II 70 kDa subunit|Ku p70
- CTC box binding factor 75 kDa subunit|Kup70|ATP dependent DNA helicase II 70 kDa subunit|DNA repair protein XRCC6|XRCC 6|Ku 70|Ku autoantigen, 70kDa|Thyroid-lupus autoantigen|70 kDa subunit of Ku antigen|Ku autoantigen p70 subunit|XRCC6_HUMAN|TLAA|X-ray repair complementing defective repair in Chinese hamster cells 6|X ray repair complementing defective repair in Chinese hamster cells 6|Thyroid autoantigen 70kD (Ku antigen)|Ku70 DNA binding component of DNA-dependent proteinkinase complex (thyroid autoantigen 70 kDa|Thyroid lupus autoantigen|Xrcc6|CTC75|CTCBF|X-ray repair cross-complementing protein 6|ATP dependent DNA helicase 2 subunit 1|Ku70|CTC box-binding factor 75 kDa subunit|Thyroid lupus autoantigen p70|ATP-dependent DNA helicase 2 subunit 1|ML8|5''-dRP lyase Ku70|Lupus Ku autoantigen protein p70|Thyroid autoantigen|G22P1|5''-deoxyribose-5-phosphate lyase Ku70|ATP-dependent DNA helicase II 70 kDa subunit|Ku p70
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P12956
- P12956
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthesized peptide derived from the Internal region of Human Ku-70.
- Synthesized peptide derived from the Internal region of Human Ku-70.
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together