Anti-HDAC4 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, IHC:1:10-1:50
Type:
Antigen: HDAC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77714-924
- 77714-925
- Antigen Symbol:
- HDAC4
- HDAC4
- Antigen Synonyms:
- HA6116|AHO3|EC 3.5.1.98|BDMR|Histone deacetylase 4|HDAC4_HUMAN|HDAC4|HDACA|HD 4|HDAC 4|Histone Deacetylase A|HDAC A|HD4|KIAA0288
- HA6116|AHO3|EC 3.5.1.98|BDMR|Histone deacetylase 4|HDAC4_HUMAN|HDAC4|HDACA|HD 4|HDAC 4|Histone Deacetylase A|HDAC A|HD4|KIAA0288
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P56524
- P56524
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human HDAC4
- Synthetic peptide of Human HDAC4
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together