Anti-CXCR1 Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:2000, WB:1:200-1:1000
Type:
Antigen: CXCR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77714-922
- 77714-923
- Antigen Symbol:
- CXCR1
- CXCR1
- Antigen Synonyms:
- Chemokine receptor 1|CD181 antigen|IL8 receptor type 1|High affinity interleukin 8 receptor A|Chemokine (C X C) receptor 1|CCCKR1|IL 8R A|IL8 receptor|CXCR-1|C C|CD181|Interleukin 8 receptor alpha|IL8RBA|IL-8R A|IL8RA|IL-8 receptor type 1|C-X-C chemokine receptor type 1|High affinity interleukin-8 receptor A|CKR 1|CXCR1|C C CKR 1|CXCR 1|CXCR1_HUMAN|Interleukin 8 receptor type 1|CD128|CDw128a|Chemokine (C X C motif) receptor 1|IL8R1|Interleukin 8 receptor type A|CMKAR1|CXC-R1|CD 181|IL 8 receptor type 1
- Chemokine receptor 1|CD181 antigen|IL8 receptor type 1|High affinity interleukin 8 receptor A|Chemokine (C X C) receptor 1|CCCKR1|IL 8R A|IL8 receptor|CXCR-1|C C|CD181|Interleukin 8 receptor alpha|IL8RBA|IL-8R A|IL8RA|IL-8 receptor type 1|C-X-C chemokine receptor type 1|High affinity interleukin-8 receptor A|CKR 1|CXCR1|C C CKR 1|CXCR 1|CXCR1_HUMAN|Interleukin 8 receptor type 1|CD128|CDw128a|Chemokine (C X C motif) receptor 1|IL8R1|Interleukin 8 receptor type A|CMKAR1|CXC-R1|CD 181|IL 8 receptor type 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P25024
- P25024
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human CXCR1
- Synthetic peptide of Human CXCR1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together