Anti-SLC16A14 Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:2000, WB:1:200-1:1000
Type:
Antigen: SLC16A14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77714-861
- 77714-862
- Antigen Symbol:
- SLC16A14
- SLC16A14
- Antigen Synonyms:
- OTTHUMP00000204390|OTTHUMP00000204389|Solute carrier family 16, member 14 (monocarboxylic acid transporter 14)|MCT 14|Solute carrier family 16 (monocarboxylic acid transporters), member 14|FLJ30794|MCT14|Solute carrier family 16 member 14|MOT14_HUMAN|Monocarboxylate transporter 14|Slc16a14
- OTTHUMP00000204390|OTTHUMP00000204389|Solute carrier family 16, member 14 (monocarboxylic acid transporter 14)|MCT 14|Solute carrier family 16 (monocarboxylic acid transporters), member 14|FLJ30794|MCT14|Solute carrier family 16 member 14|MOT14_HUMAN|Monocarboxylate transporter 14|Slc16a14
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7RTX9
- Q7RTX9
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human SLC16A14
- Synthetic peptide of Human SLC16A14
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together