Anti-DHCR24 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, WB:1:200-1:1000, IHC:1:30-1:150
Type:
Antigen: DHCR24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77714-855
- 77714-854
- Antigen Symbol:
- DHCR24
- DHCR24
- Antigen Synonyms:
- 24-dehydrocholesterol reductase|3-beta-hydroxysterol delta-24-reductase|Nbla03646|KIAA0018|DCE|SELADIN1|Selective Alzheimer's Disease Indicator 1|Diminuto/dwarf1 homolog|Seladin-1|Dhcr24|Delta(24)-sterol reductase|Diminuto dwarf1 homolog|DHC24_HUMAN|3 beta hydroxysterol delta 24 reductase|24 dehydrocholesterol reductase|Desmosterol to cholesterol enzyme|Selective AD indicator 1
- 24-dehydrocholesterol reductase|3-beta-hydroxysterol delta-24-reductase|Nbla03646|KIAA0018|DCE|SELADIN1|Selective Alzheimer's Disease Indicator 1|Diminuto/dwarf1 homolog|Seladin-1|Dhcr24|Delta(24)-sterol reductase|Diminuto dwarf1 homolog|DHC24_HUMAN|3 beta hydroxysterol delta 24 reductase|24 dehydrocholesterol reductase|Desmosterol to cholesterol enzyme|Selective AD indicator 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15392
- Q15392
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human DHCR24
- Fusion protein of Human DHCR24
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together