Anti-EXTL3 Rabbit Polyclonal Antibody
Catalog # 77688-948
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:EXTL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:O43909
- Antigen Synonyms:REGR|EXTR1|Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferas|botv|RPR|EXTL3_HUMAN|Multiple exostosis-like protein 3|Putative tumor suppressor protein EXTL3|Exostoses (multiple)-like 3|EXTL1L|Hereditary multiple exostoses gene isolog|KIAA0519|Exostosin-like 3|Exostoses-like 3|EXT-related protein 1|Reg receptor|DKFZp686C2342|EXTL3|REG|Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Exostosin-like 3 protein (2-245AA)
- Purification:>95%, Protein G purified
- Cat. No.:77688-948
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200, IF:1:50-1:200
Type:
Antigen: EXTL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human