Anti-PRKACA/PRKACB/PRKACG (phospho-T198) Rabbit Polyclonal Antibody
Catalog # 77703-800
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:PRKACA/PRKACB/PRKACG (phospho-T198)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:P17612/P22694/P22612
- Antigen Synonyms:PKA C-beta|cAMP-dependent protein kinase catalytic subunit gamma|PRKACG|PKA C-alpha|PRKACB|PRKACA|PKACA|cAMP-dependent protein kinase catalytic subunit alpha|cAMP-dependent protein kinase catalytic subunit beta|PKA C-gamma
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human PKAα/β/γ cat around the phosphorylation site of T198.
- Purification:Antigen affinity purified
- Cat. No.:77703-800
- Packaging:Vial
Specifications
About this item
Type:
Antigen: PRKACA/PRKACB/PRKACG (phospho-T198)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat