Anti-ATP5G2 Rabbit Polyclonal Antibody
Catalog # 77703-625
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ATP5G2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q06055
- Antigen Synonyms:ATP synthase proteolipid P2|Mitochondrial ATP synthase subunit C subunit 9 isoform 2|subunit c (subunit 9)|subunit c|ATPase protein 9|ATP synthase F(0) complex subunit C2|ATP synthase proton-transporting mitochondrial F(0) complex subunit C2|mitochondrial Fo complex|ATP synthase H+ transporting mitochondrial F0 complex subunit C2|ATPase subunit c|mitochondrial|ATP synthase|Mitochondrial ATP synthase|OTTHUMP00000164642|subunit C2 (subunit 9)|ATP synthase lipid binding protein mitochondrial|mitochondrial F0 complex|isoform 2|subunit 9|ATP synthase H+ transporting mitochondrial F0 complex subunit c subunit 9 isoform 2|ATP synthase H+ transporting mitochondrial F0 complex subunit C2 subunit 9|AT5G2_HUMAN|ATP5A|ATP synthase c subunit|H+ transporting|ATP5G2|ATP synthase lipid-binding protein|PSEC0033
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human ATP5G2.
- Purification:Antigen affinity purified
- Cat. No.:77703-625
- Packaging:Vial
Specifications
About this item
IHC:1:100-1:300, ELISA:1:40000
Type:
Antigen: ATP5G2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human