Anti-ZBTB25 Rabbit Polyclonal Antibody
Catalog # 77688-889
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ZBTB25
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P24278
- Antigen Synonyms:Zinc finger protein KUP|ZBTB25|Zinc finger and BTB domain-containing protein 25|ZBT25_HUMAN|Zinc finger protein 46
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Zinc finger and BTB domain-containing protein 25 protein (266-435AA)
- Purification:Antigen affinity purified
- Cat. No.:77688-889
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: ZBTB25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human