Anti-TRAF6 Rabbit Polyclonal Antibody
Catalog # 77688-828
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:TRAF6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q9Y4K3
- Antigen Synonyms:RING finger protein 85|Interleukin-1 signal transducer|MGC3310|OTTHUMP00000232772|TNF receptor-associated factor 6|RNF85|TRAF6_HUMAN|Traf6|OTTHUMP00000232773|TRAF 6|RNF 85|MGC 3310|E3 ubiquitin protein ligase|MGC:3310|E3 ubiquitin-protein ligase TRAF6|Interleukin 1 signal transducer|TNF receptor associated factor 6
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human TNF receptor-associated factor 6 protein (1-522AA)
- Purification:>95%, Protein G purified
- Cat. No.:77688-828
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TRAF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human