Anti-PAK1/PAK2/PAK3 Rabbit Polyclonal Antibody
77703-540
New Product
- Antigen Symbol:PAK1/PAK2/PAK3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q13153/Q13177/O75914
- Antigen Synonyms:S6/H4 kinase|PAK-2|Serine/threonine-p|p65-PAK|p58|Serine/threonine-protein kinase PAK 2|Gamma-PAK|Alpha-PAK|p21-activated kinase 1|p21-activated kinase 2|PAK3|PAK65|PAK-1|PAK1|OPHN3|PAK2|Serine/threonine-protein kinase PAK 1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human PAKα/β/γ around the non-phosphorylation site of T423/402/421.
- Purification:Antigen affinity purified
- Cat. No.:77703-540
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:20000
Type:
Antigen: PAK1/PAK2/PAK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat