Anti-RPL24 Rabbit Polyclonal Antibody
Catalog # 77688-596
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:RPL24
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Gene ID:P83731
- Antigen Synonyms:Ribosomal protein L30|60S ribosomal protein L24|L24|RPL24|60S ribosomal protein L30|Ribosomal protein L24|RL24_HUMAN|RPL 24
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human 60S ribosomal protein L24 protein (1-157AA)
- Purification:>95%, Protein G purified
- Cat. No.:77688-596
- Packaging:Vial
Specifications
About this item
Type:
Antigen: RPL24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human