Anti-PRKAA2 (phospho-S491) HEK293F cells Recombinant Monoclonal Antibody [clone: 3H10]
Catalog # 77691-373
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Symbol:PRKAA2 (phospho-S491)
- Clonality:Recombinant Monoclonal
- Clone:3H10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:HEK293F cells
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P54646
- Antigen Synonyms:AMPK|AMPK2|Acetyl-CoA carboxylase kinase|PRKAA2|5'-AMP-activated protein kinase catalytic subunit alpha-2
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:A synthesized peptide derived from Human Phospho-PRKAA2 (S491)
- Purification:Affinity purified
- Cat. No.:77691-373
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:5000
Type:
Antigen: PRKAA2 (phospho-S491)
Clonality: Recombinant Monoclonal
Clone: 3H10
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: Rabbit IgG
Reactivity: Human