To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Symbol:Mono-methyl-Histone H3.1(R17)Antibody
- Clonality:Recombinant Monoclonal
- Clone:3E10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:HEK293F cells
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P68431
- Antigen Synonyms:H3FA|Histone H3/l|HIST1H3J|H3FJ|HIST1H3E|HIST1H3G|HIST1H3B|Histone H3/f|H3FI|HIST1H3D|HIST1H3C|HIST1H3I|HIST1H3F|Histone H3/b|Histone H3/d|AND|Histone H3.1|Histone H3/i|H3FH|H3FD|Histone H3/a|Histone H3/k|H3FF|HIST1H3H|H3FC|H3FB|Histone H3/j|H3FL|Histone H3/c|Histone H3/h|H3FK|HIST1H3A
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide
- Purification:Affinity purified
- Cat. No.:77691-285
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:2000, ICC:1:50-1:500, IF:1:30-1:200
Type:
Antigen: Mono-methyl-Histone H3.1(R17)Antibody
Clonality: Recombinant Monoclonal
Clone: 3E10
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: Rabbit IgG
Reactivity: Human