Anti-ACER1 Rabbit Polyclonal Antibody
Catalog # 77705-670
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ACER1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q8TDN7
- Antigen Synonyms:N acylsphingosine amidohydrolase 3|Acylsphingosine deacylase 3|ASAH 3|Acer1|N acylsphingosine amidohydrolase 3 alkaline ceramidase|ALKCDase1|AlkCDase 1|N acylsphingosine amidohydrolase (alkaline ceramidase) 3|N-acylsphingosine amidohydrolase 3|Alkaline ceramidase 1|MGC138327|ACER1_HUMAN|MGC138329|Alkaline ceramidase|Alkaline CDase 1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human ASAH3. at AA rangle: 100-180
- Purification:Antigen affinity purified
- Cat. No.:77705-670
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:10000
Type:
Antigen: ACER1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human