Anti-RCE1 Rabbit Polyclonal Antibody
Catalog # 77691-144
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:RCE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9Y256
- Antigen Synonyms:FACE-2|EC 3.4.22.-|RCE1A|hRCE1|RCE1|FACE2|Prenyl protein-specific endoprotease 2|Farnesylated proteins-converting enzyme 2|RCE1BCAAX prenyl protease 2|RCE1 homolog
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human CAAX prenyl protease 2 protein (133-185AA)
- Purification:>95%, Protein G purified
- Cat. No.:77691-144
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:5000
Type:
Antigen: RCE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human