Anti-SERPINA10 Rabbit Polyclonal Antibody
Catalog # 77691-084
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:SERPINA10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9UK55
- Antigen Synonyms:SERPINA 10|PZ-dependent protease inhibitor|Protein Z-dependent protease inhibitor|Serpin A10|PZ dependent protease inhibitor|ZPI|serine (or cysteine) proteinase inhibitor clade A (alpha 1 antiproteinase antitrypsin) member 10|PZI|SERPINA10|Protein Z dependent protease inhibitor precursor|Serpin peptidase inhibitor clade A (alpha 1 antiproteinase antitrypsin) member 10|ZPI_HUMAN|Protein Z dependent protease inhibitor
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Protein Z-dependent protease inhibitor protein (165-444AA)
- Purification:Antigen affinity purified
- Cat. No.:77691-084
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200
Type:
Antigen: SERPINA10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse