Anti-CALCOCO1 Rabbit Polyclonal Antibody
77690-846
New Product
- Antigen Symbol:CALCOCO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9P1Z2
- Antigen Synonyms:KIAA1536|Calphoglin|PP13275|Coiled-coil coactivator protein|CALCOCO1|UNQ2436/PRO4996Calcium-binding and coiled-coil domain-containing protein 1|Sarcoma antigen NY-SAR-3
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Calcium-binding and coiled-coil domain-containing protein 1 protein (462-691AA)
- Purification:Antigen affinity purified
- Cat. No.:77690-846
- Packaging:Vial
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: CALCOCO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human