Anti-MSL2 Rabbit Polyclonal Antibody
Catalog # 77690-822
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MSL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9HCI7
- Antigen Synonyms:MSL2L1|Male-specific lethal-2 homolog 1|MSL2_HUMAN|Male specific lethal 2 homolog (Drosophila)|Male specific lethal 2 homolog 1|Ring finger protein 184|Male-specific lethal 2 homolog|Male-specific lethal 2-like 1|MSL2-like 1|MSL2|Male specific lethal 2 like 1|Male specific lethal 2 homolog|MSL-2
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human E3 ubiquitin-protein ligase MSL2 protein (341-473AA)
- Purification:>95%, Protein G purified
- Cat. No.:77690-822
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:5000
Type:
Antigen: MSL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human