Anti-MOB1A Rabbit Polyclonal Antibody
77690-753
New Product
- Antigen Symbol:MOB1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9H8S9
- Antigen Synonyms:FLJ10788|MOB4B|Mob1 homolog 1B|C2orf6|Mps one binder kinase activator-like 1B|Chromosome 2 open reading frame 6|Mob1 alpha|MOB kinase activator 1A|MOB1|MATS1|Mob1A|MOBK1B|MOL1B_HUMAN|Mps one binder kinase activator like 1B|FLJ11595|Mps One Binder kinase activator like 1B (yeast)|B1|MOB1 Mps One Binder kinase activator like 1B|Mobkl1b|MOB1 Mps One Binder homolog A|Mob4B protein|Protein Mob4B
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human MOB kinase activator 1A protein (2-216AA)
- Purification:>95%, Protein G purified
- Cat. No.:77690-753
- Packaging:Vial
Recommended dilution: WB:1:2000-1:5000, IHC:1:100-1:1000, IF:1:200-1:500, IP1:200-1:2000
Type:
Antigen: MOB1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human