Anti-SLC25A23 Rabbit Polyclonal Antibody
77690-736
New Product
- Antigen Symbol:SLC25A23
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9BV35
- Antigen Synonyms:APC2|Solute carrier family 25 member 23|SLC25A23|Mitochondrial ATP-Mg/Pi carrier protein 2|SCAMC3Calcium-binding mitochondrial carrier protein SCaMC-3|Mitochondrial Ca(2+)-dependent solute carrier protein 2|MCSC2|Small calcium-binding mitochondrial carrier protein 3
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Calcium-binding mitochondrial carrier protein SCaMC-3 protein (1-188AA)
- Purification:>95%, Protein G purified
- Cat. No.:77690-736
- Packaging:Vial
Recommended dilution: WB:1:500-1:5000
Type:
Antigen: SLC25A23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse