Anti-CYP4F12 Rabbit Polyclonal Antibody
77690-643
New Product
- Antigen Symbol:CYP4F12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9HCS2
- Antigen Synonyms:Cytochrome P450|CYP4F12|Cytochrome P450 4F12|CYPIVF12|CP4FC_HUMAN|subfamily F|polypeptide 12|family 4
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cytochrome P450 4F12 protein (265-524AA)
- Purification:Antigen affinity purified
- Cat. No.:77690-643
- Packaging:Vial
Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200
Type:
Antigen: CYP4F12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat