Anti-PPIL3 Rabbit Polyclonal Antibody
Catalog # 77690-631
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:PPIL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9H2H8
- Antigen Synonyms:Peptidylprolyl isomerase (cyclophilin) like 3|Cyclophilin J|Cyclophilin like 3|Cyp10l|Peptidylprolyl cis trans isomerase like protein 3|CyPJ|2310076N22Rik|PPIase like protein 3|Rotamase|Cyclophilin like protein 3|Peptidylprolyl isomerase like 3|Ppil3|Ccyclophilin like protein|Cyclophilin-like protein PPIL3|2510026K04Rik|MGC105285|Cyclophilin like protein PPIL3|Rotamase PPIL3|Peptidyl-prolyl cis-trans isomerase-like 3|PPIase|PPIL3_HUMAN|Peptidyl prolyl cis trans isomerase like 3
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Peptidyl-prolyl cis-trans isomerase-like 3 protein (2-161AA)
- Purification:>95%, Protein G purified
- Cat. No.:77690-631
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: PPIL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human