Anti-YIPF3 Rabbit Polyclonal Antibody
Catalog # 77690-626
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:YIPF3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9GZM5
- Antigen Synonyms:C6orf109|FinGER3|YIP1 family member 3|Protein YIPF3|dJ337H4.3|KLIP1|Yip1 domain family member 3|YIPF3_HUMAN|Natural killer cell-specific antigen KLIP1|Yipf3|Killer lineage protein 1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Protein YIPF3 protein (2-148AA)
- Purification:>95%, Protein G purified
- Cat. No.:77690-626
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200
Type:
Antigen: YIPF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse