Anti-DUSP26 Rabbit Polyclonal Antibody
Catalog # 77690-614
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:DUSP26
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q9BV47
- Antigen Synonyms:NATA1 protein|NATA1|DSP-4|MGC2627|MAP kinase phosphatase 8|Low molecular mass dual specificity phosphatase 4|DUSP24|DUSP26|MKP-8|Dual specificity phosphatase SKRP3|Hypothetical protein FLJ31142|Dual specificity phosphatase 26 (putative)|Dual specificity phosphatase 26|Low-molecular-mass dual-specificity phosphatase 4|MGC1136|Mitogen activated protein kinase phosphatase 8|LDP-4|Mitogen-activated protein kinase phosphatase 8|DUS26_HUMAN|Novel amplified gene in thyroid anaplastic cancer|Dual specificity protein phosphatase 26|MKP8|SKRP3|LDP 4
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Dual specificity protein phosphatase 26 protein (1-211AA)
- Purification:>95%, Protein G purified
- Cat. No.:77690-614
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC:1:20-1:200, IF:1:50-1:500
Type:
Antigen: DUSP26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human