Anti-PITX2 Rabbit Polyclonal Antibody
77690-575
New Product
- Antigen Symbol:PITX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q99697
- Antigen Synonyms:IDG 2|ALL1 responsive protein ARP1|IDG2|Rieg bicoid related homeobox transcription factor 1|All1 responsive gene 1|PITX 2|IHG 2|RIEG1|RIEG bicoid-related homeobox transcription factor|ARP 1|IHG2|IGDS2|RIEG|IRID 2|pitx2|Otlx 2|PITX2_HUMAN|IGDS 2|Paired like homeodomain transcription factor 2|RS|Pituitary homeo box 2|Brx 1|Brx1|RIEG bicoid related homeobox transcription factor|IRID2|IGDS|Pituitary homeobox 2|Solurshin|ARP1|RIEG 1|PTX 2|RGS|PTX2|ALL1-responsive protein ARP1|Paired-like homeodomain transcription factor 2|Otlx2|MGC111022|MGC20144|Homeobox protein PITX2
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Pituitary homeobox 2 protein (40-154AA)
- Purification:>95%, Protein G purified
- Cat. No.:77690-575
- Packaging:Vial
Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200
Type:
Antigen: PITX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human